Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022400.1 | NP_071795.1 | 4 - 5 | 422 | 82 | |
XM_006229149.1 | XP_006229211.1 | 4 - 5 | 599 | 82 | ||
GenBank mRNA | U68417.1 | - | 4 - 5 | 422 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |