Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107503.1 | NP_001100973.1 | 11 - 12 | 2639 | 98 | |
XM_006228828.2 | XP_006228890.1 | 13 - 14 | 2620 | 98 | ||
XM_008759152.1 | XP_008757374.1 | 12 - 13 | 2693 | 98 | ||
XM_008759153.1 | XP_008757375.1 | 12 - 13 | 2454 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |