Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164060.1 | NP_001157532.1 | 7 - 8 | 1193 | 70 | |
NM_053517.2 | NP_445969.2 | 8 - 9 | 760 | 70 | ||
GenBank mRNA | BC088298.1 | - | 8 - 9 | 760 | 7323045 | 70 |
CB813924.1 | - | 1 - 2 | 40 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |