Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232539.2 | XP_006232601.1 | 4 - 5 | 620 | 60 | |
XM_006232540.2 | XP_006232602.1 | 5 - 6 | 762 | 60 | ||
XM_006232541.2 | XP_006232603.1 | 6 - 7 | 829 | 60 | ||
XM_006232543.1 | XP_006232605.1 | 4 - 5 | 372 | 60 | ||
XM_008761110.1 | XP_008759332.1 | 4 - 5 | 376 | 60 | ||
GenBank mRNA | FQ222383.1 | - | 4 - 5 | 392 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |