Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133565.1 | NP_598249.1 | 3 - 4 | 267 | 79 | |
GenBank mRNA | FQ217639.1 | - | 3 - 4 | 262 | 79 | |
FQ217678.1 | - | 3 - 4 | 261 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |