Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015005.2 | NP_001015005.2 | 8 - 9 | 909 | 72 | |
XM_006237518.1 | XP_006237580.1 | 9 - 10 | 966 | 72 | ||
GenBank mRNA | BC091696.1 | - | 8 - 9 | 902 | 7301732 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |