Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009536.1 | NP_001009536.1 | 2 - 3 | 748 | 73 | |
GenBank mRNA | BC061749.1 | - | 2 - 3 | 748 | 5599464 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |