Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107012.1 | NP_001100482.1 | 8 - 9 | 1451 | 146 | |
XM_006246704.2 | XP_006246766.1 | 9 - 10 | 2909 | 146 | ||
XM_006246705.2 | XP_006246767.1 | 10 - 11 | 1984 | 146 | ||
XM_006246707.2 | XP_006246769.1 | 10 - 11 | 1654 | 146 | ||
XM_006246709.2 | XP_006246771.1 | 9 - 10 | 1831 | 146 | ||
XM_008767827.1 | XP_008766049.1 | 9 - 10 | 2360 | 146 | ||
XM_008767828.1 | XP_008766050.1 | 10 - 11 | 1748 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |