Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053665.1 | NP_446117.1 | 2 - 3 | 1696 | 131 | |
XM_006247067.2 | XP_006247129.1 | 3 - 4 | 1869 | 131 | ||
XM_006247068.1 | XP_006247130.1 | 3 - 4 | 1865 | 131 | ||
XM_008767930.1 | XP_008766152.1 | 3 - 4 | 1924 | 131 | ||
GenBank mRNA | AF068202.1 | - | 2 - 3 | 1696 | 131 | |
BC081753.1 | - | 4 - 5 | 1948 | 131 | ||
BC097298.1 | - | 5 - 6 | 2040 | 131 | ||
BC129090.1 | - | 5 - 6 | 2019 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |