Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011892.1 | NP_001011892.1 | 6 - 7 | 552 | 131 | |
XM_006246675.1 | XP_006246737.1 | 6 - 7 | 697 | 131 | ||
GenBank mRNA | BC079362.1 | - | 6 - 7 | 552 | 7109243 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |