Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191591.1 | NP_001178520.1 | 1 - 2 | 81 | 124 | |
XM_006247682.2 | XP_006247744.1 | 1 - 2 | 92 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Inventoried Probe spans exons |