Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053766.1 | NP_446218.1 | 5 - 6 | 556 | 88 | |
XM_008770840.1 | XP_008769062.1 | 2 - 3 | 1072 | 88 | ||
XM_008770841.1 | XP_008769063.1 | 3 - 4 | 575 | 88 | ||
GenBank mRNA | BC089052.1 | - | 5 - 6 | 556 | 7307299 | 88 |
FQ226684.1 | - | 4 - 5 | 338 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |