Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105789.1 | NP_001099259.1 | 9 - 10 | 1793 | 99 | |
XM_008767788.1 | XP_008766010.1 | 8 - 9 | 3240 | 99 | ||
XM_008767789.1 | XP_008766011.1 | 8 - 9 | 3800 | 99 | ||
XM_008767790.1 | XP_008766012.1 | 7 - 8 | 2129 | 99 | ||
XM_008767791.1 | XP_008766013.1 | 8 - 9 | 1977 | 99 | ||
XM_008767792.1 | XP_008766014.1 | 8 - 9 | 1975 | 99 | ||
XM_008767793.1 | XP_008766015.1 | 7 - 8 | 2131 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |