Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013024.2 | NP_037156.2 | 3 - 4 | 372 | 73 | |
XM_008767548.1 | XP_008765770.1 | 2 - 3 | 178 | 73 | ||
GenBank mRNA | CA339008.1 | - | 3 - 4 | 372 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |