Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twistnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108707.1 | NP_001102177.1 | 1 - 2 | 354 | 95 | |
GenBank mRNA | FQ230315.1 | - | 1 - 2 | 269 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |