Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768149.1 | XP_008766371.1 | 13 - 14 | 1246 | 60 | |
GenBank mRNA | FQ210246.1 | - | 13 - 14 | 1214 | 60 | |
FQ217168.1 | - | 13 - 14 | 1210 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |