Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107075.1 | NP_001100545.1 | 2 - 3 | 314 | 123 | |
XM_006247890.2 | XP_006247952.1 | 2 - 3 | 711 | 123 | ||
XM_006247891.2 | XP_006247953.1 | 2 - 3 | 711 | 123 | ||
GenBank mRNA | BC168919.1 | - | 1 - 2 | 179 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |