Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105800.1 | NP_001099270.1 | 7 - 8 | 1903 | 116 | |
XM_006246661.2 | XP_006246723.1 | 6 - 7 | 1418 | 116 | ||
XM_006246662.2 | XP_006246724.1 | 6 - 7 | 1405 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |