Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013149.2 | NP_037281.2 | 6 - 7 | 697 | 87 | |
GenBank mRNA | AF082124.1 | - | 6 - 7 | 726 | 87 | |
AF082125.1 | - | 6 - 7 | 726 | 87 | ||
AF082126.1 | - | 6 - 7 | 726 | 87 | ||
AJ821851.1 | - | 6 - 7 | 670 | 87 | ||
AM902281.1 | - | 6 - 7 | 697 | 87 | ||
AM902282.1 | - | 6 - 7 | 697 | 87 | ||
AM902283.1 | - | 6 - 7 | 697 | 87 | ||
AM902284.1 | - | 6 - 7 | 697 | 87 | ||
AM902285.1 | - | 6 - 7 | 697 | 87 | ||
AM902286.1 | - | 6 - 7 | 697 | 87 | ||
U09000.1 | - | 6 - 7 | 697 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |