Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108024.1 | NP_001101494.1 | 1 - 2 | 289 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |