Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240244.2 | XP_006240306.1 | 33 - 34 | 6869 | 83 | |
XM_008764770.1 | XP_008762992.1 | 33 - 34 | 6869 | 83 | ||
XM_008764771.1 | XP_008762993.1 | 34 - 35 | 6981 | 83 | ||
XM_008764772.1 | XP_008762994.1 | 34 - 35 | 6963 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |