Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108034.1 | NP_001101504.1 | 3 - 4 | 826 | 124 | |
XM_006240258.2 | XP_006240320.1 | 4 - 5 | 1395 | 124 | ||
XM_008764773.1 | XP_008762995.1 | 4 - 5 | 999 | 124 | ||
XM_008764774.1 | XP_008762996.1 | 4 - 5 | 763 | 124 | ||
GenBank mRNA | BC168247.1 | - | 4 - 5 | 884 | 5597968 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |