Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013940.1 | NP_001013962.1 | 3 - 4 | 238 | 120 | |
XM_006239695.2 | XP_006239757.1 | 3 - 4 | 293 | 120 | ||
GenBank mRNA | BC079201.1 | - | 3 - 4 | 238 | 7131728 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |