Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727893.3 | XP_002727939.1 | 7 - 8 | 802 | 78 | |
XM_006248441.2 | XP_006248503.1 | 3 - 4 | 1316 | 78 | ||
XM_008768770.1 | XP_008766992.1 | 3 - 4 | 1231 | 78 | ||
XM_008768771.1 | XP_008766993.1 | 3 - 4 | 380 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |