Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169146.1 | NP_001162617.1 | 1 - 2 | 127 | 80 | |
XM_006228612.2 | XP_006228674.1 | 1 - 2 | 124 | 80 | ||
GenBank mRNA | BC064030.1 | - | 101 | 80 | ||
BC084715.1 | - | 1 - 2 | 96 | 80 | ||
BC159438.1 | - | 1 - 2 | 98 | 80 | ||
FQ211832.1 | - | 1 - 2 | 108 | 80 | ||
FQ221089.1 | - | 1 - 2 | 115 | 80 | ||
FQ221637.1 | - | 1 - 2 | 109 | 80 | ||
FQ221660.1 | - | 1 - 2 | 108 | 80 | ||
FQ221692.1 | - | 1 - 2 | 106 | 80 | ||
FQ222913.1 | - | 1 - 2 | 108 | 80 | ||
FQ223100.1 | - | 1 - 2 | 109 | 80 | ||
FQ223384.1 | - | 1 - 2 | 108 | 80 | ||
FQ223396.1 | - | 1 - 2 | 109 | 80 | ||
FQ224101.1 | - | 1 - 2 | 108 | 80 | ||
FQ228433.1 | - | 1 - 2 | 109 | 80 | ||
FQ229060.1 | - | 1 - 2 | 61 | 80 | ||
X17665.1 | - | 1 - 2 | 107 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |