Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004133.1 | NP_001004133.1 | 2 - 3 | 720 | 74 | |
NM_133440.1 | NP_597684.1 | 2 - 3 | 826 | 74 | ||
NM_133441.1 | NP_597685.1 | 7 - 8 | 1320 | 74 | ||
NM_201350.1 | NP_958738.1 | 16 - 17 | 2169 | 74 | ||
XM_008760068.1 | XP_008758290.1 | 15 - 16 | 2080 | 74 | ||
GenBank mRNA | AF060116.1 | - | 2 - 3 | 720 | 74 | |
AF141903.1 | - | 6 - 7 | 812 | 74 | ||
AF159048.1 | - | 6 - 7 | 830 | 74 | ||
AJ131899.1 | - | 2 - 3 | 826 | 74 | ||
AJ249562.1 | - | 7 - 8 | 1320 | 74 | ||
AY298755.1 | - | 16 - 17 | 2169 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |