Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130039.1 | NP_001123511.1 | 3 - 4 | 892 | 98 | |
XM_006230443.2 | XP_006230505.1 | 3 - 4 | 1457 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |