Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009681.1 | NP_001009681.1 | 2 - 3 | 493 | 56 | |
GenBank mRNA | AY227756.1 | - | 2 - 3 | 493 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |