Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728259.4 | XP_002728305.2 | 8 | 3392 | 115 | |
XM_006252082.2 | XP_006252144.1 | 7 | 3296 | 115 | ||
XM_006252083.2 | XP_006252145.1 | 7 | 3232 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |