Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005871.1 | NP_001005871.1 | 16 - 17 | 2980 | 72 | |
XM_008769446.1 | XP_008767668.1 | 17 - 18 | 3363 | 72 | ||
XM_008769447.1 | XP_008767669.1 | 17 - 18 | 4000 | 72 | ||
XM_008769448.1 | XP_008767670.1 | 16 - 17 | 3327 | 72 | ||
XM_008769449.1 | XP_008767671.1 | 17 - 18 | 3364 | 72 | ||
XM_008769450.1 | XP_008767672.1 | 16 - 17 | 3328 | 72 | ||
GenBank mRNA | U15408.1 | - | 16 - 17 | 2980 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |