Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191889.1 | NP_001178818.1 | 9 - 10 | 1965 | 69 | |
XM_006234247.2 | XP_006234309.1 | 9 - 10 | 2260 | 69 | ||
XM_006234248.2 | XP_006234310.1 | 9 - 10 | 1906 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |