Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109594.1 | NP_001103064.1 | 9 - 10 | 1160 | 90 | |
XM_008762035.1 | XP_008760257.1 | 8 - 9 | 1188 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |