Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Knstrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004264.1 | NP_001004264.1 | 6 - 7 | 749 | 99 | |
XM_006234757.2 | XP_006234819.1 | 6 - 7 | 785 | 99 | ||
XM_006234758.2 | XP_006234820.1 | 6 - 7 | 606 | 99 | ||
GenBank mRNA | BC079438.1 | - | 6 - 7 | 749 | 7115344 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |