Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031773.1 | NP_113961.1 | 13 - 14 | 2328 | 65 | |
GenBank mRNA | AF025424.1 | - | 13 - 14 | 2328 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |