Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254331.2 | XP_006254393.1 | 3 - 4 | 1891 | 64 | |
XM_006254332.2 | XP_006254394.1 | 3 - 4 | 1077 | 64 | ||
XM_008771904.1 | XP_008770126.1 | 6 - 7 | 1828 | 64 | ||
XM_008771905.1 | XP_008770127.1 | 6 - 7 | 1828 | 64 | ||
XM_008771908.1 | XP_008770130.1 | 6 - 7 | 1828 | 64 | ||
XM_008771909.1 | XP_008770131.1 | 6 - 7 | 1450 | 64 | ||
XM_008771910.1 | XP_008770132.1 | 6 - 7 | 1830 | 64 | ||
XM_008771912.1 | XP_008770134.1 | 6 - 7 | 1830 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |