Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177421.3 | NP_803156.2 | 6 - 7 | 1291 | 113 | |
XM_006251965.1 | XP_006252027.1 | 6 - 7 | 1929 | 113 | ||
GenBank mRNA | AB040056.1 | - | 6 - 7 | 1298 | 113 | |
BC099072.1 | - | 6 - 7 | 1291 | 7312276 | 113 | |
FQ212273.1 | - | 6 - 7 | 1293 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |