Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017257.2 | NP_058953.1 | 9 - 10 | 551 | 60 | |
GenBank mRNA | BC058486.1 | - | 9 - 10 | 589 | 6918985 | 60 |
BC097331.1 | - | 9 - 10 | 551 | 7389180 | 60 | |
D45250.1 | - | 9 - 10 | 601 | 60 | ||
FQ226805.1 | - | 9 - 10 | 582 | 60 | ||
FQ227049.1 | - | 9 - 10 | 595 | 60 | ||
FQ232840.1 | - | 9 - 10 | 581 | 60 | ||
FQ233763.1 | - | 9 - 10 | 577 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |