Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017264.3 | NP_058960.2 | 5 - 6 | 402 | 127 | |
GenBank mRNA | BC060574.1 | - | 5 - 6 | 406 | 6890794 | 127 |
D45249.1 | - | 5 - 6 | 326 | 127 | ||
FQ216423.1 | - | 5 - 6 | 403 | 127 | ||
FQ220175.1 | - | 5 - 6 | 351 | 127 | ||
FQ224986.1 | - | 5 - 6 | 349 | 127 | ||
FQ230622.1 | - | 5 - 6 | 127 | |||
FQ233032.1 | - | 5 - 6 | 355 | 127 | ||
FQ233635.1 | - | 5 - 6 | 355 | 127 | ||
FQ233859.1 | - | 5 - 6 | 351 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |