Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109330.1 | NP_001102800.1 | 2 - 3 | 598 | 69 | |
XM_006252007.2 | XP_006252069.1 | 2 - 3 | 688 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |