Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109505.1 | NP_001102975.1 | 4 - 5 | 389 | 137 | |
XM_006231756.2 | XP_006231818.1 | 5 - 6 | 656 | 137 | ||
XM_006231757.2 | XP_006231819.1 | 4 - 5 | 1427 | 137 | ||
XM_006231758.2 | XP_006231820.1 | 4 - 5 | 405 | 137 | ||
XM_006231759.2 | XP_006231821.1 | 4 - 5 | 425 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |