Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007648.1 | NP_001007649.1 | 4 - 5 | 1086 | 68 | |
XM_006237341.2 | XP_006237403.1 | 4 - 5 | 670 | 68 | ||
XM_006237342.2 | XP_006237404.1 | 3 - 4 | 1103 | 68 | ||
XM_008763281.1 | XP_008761503.1 | 4 - 5 | 808 | 68 | ||
GenBank mRNA | BC079204.1 | - | 4 - 5 | 1086 | 7131776 | 68 |
FQ209817.1 | - | 1 - 2 | 231 | 68 | ||
FQ235136.1 | - | 4 - 5 | 624 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |