Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF346902.1 | - | 1 - 2 | 122 | 63 | |
J05446.1 | - | 1 - 2 | 167 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus 1700 Probe spans exons |