Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF346902.1 | - | 3 - 4 | 490 | 99 | |
J05446.1 | - | 3 - 4 | 535 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |