Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014256.2 | NP_001014278.2 | 3 - 4 | 282 | 58 | |
GenBank mRNA | BC079366.1 | - | 3 - 4 | 282 | 7109516 | 58 |
CK481317.1 | - | 4 - 5 | 377 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |