Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107062.2 | NP_001100532.2 | 10 - 11 | 2506 | 52 | |
XM_006247686.2 | XP_006247748.1 | 10 - 11 | 2775 | 52 | ||
XM_006247687.2 | XP_006247749.1 | 10 - 11 | 2775 | 52 | ||
GenBank mRNA | BC169068.1 | - | 2504 | 52 | ||
FQ212043.1 | - | 10 - 11 | 2780 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |