Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258237.1 | NP_001245166.1 | 4 - 5 | 481 | 74 | |
GenBank mRNA | BC085122.1 | - | 4 - 5 | 481 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |