Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768134.1 | XP_008766356.1 | 10 - 11 | 1257 | 67 | |
XM_008768135.1 | XP_008766357.1 | 10 - 11 | 1260 | 67 | ||
XM_008768136.1 | XP_008766358.1 | 10 - 11 | 1256 | 67 | ||
XM_008768137.1 | XP_008766359.1 | 10 - 11 | 1256 | 67 | ||
XM_008768138.1 | XP_008766360.1 | 10 - 11 | 1256 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |