Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728858.3 | XP_002728904.1 | 1 - 2 | 1865 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |