Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap15-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105896.1 | NP_001099366.1 | 1 | 738 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |