Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240051.2 | XP_006240113.1 | 1 - 2 | 108 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |